Cytosine, 99+%
Cytosine, 99+%
Cytosine, 99+%
Thermo Scientific Chemicals

Cytosine, 99+%

CAS: 71-30-7 | C4H5N3O | 111.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 161760050
Price (USD)
80.65
Online exclusive
88.90
Save 8.25
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
80.65
Online exclusive
88.90
Save 8.25
Each
Chemical Identifiers
CAS71-30-7
IUPAC Name6-amino-1,2-dihydropyrimidin-2-one
Molecular FormulaC4H5N3O
InChI KeyOPTASPLRGRRNAP-UHFFFAOYSA-N
SMILESNC1=CC=NC(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<0.5 % (105°C, 5 h)
Appearance (Color)White to light yellow
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
HPLC>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only