3-Bromopropionyl chloride, 95%, tech.
3-Bromopropionyl chloride, 95%, tech.
3-Bromopropionyl chloride, 95%, tech.
Thermo Scientific Chemicals

3-Bromopropionyl chloride, 95%, tech.

CAS: 15486-96-1 | C3H4BrClO | 171.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 161990250
Price (USD)
79.65
Online exclusive
87.90
Save 8.25
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
79.65
Online exclusive
87.90
Save 8.25
Each
Chemical Identifiers
CAS15486-96-1
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4920 to 1.4970 (20°C, 589 nm)
GC>=94 %
Appearance (Color)Clear yellow
Appearance (Form)Liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only