Difluoroacetic acid, 98%
Difluoroacetic acid, 98%
Difluoroacetic acid, 98%
Acros Organics

Difluoroacetic acid, 98%

CAS: 381-73-7 | C2H2F2O2 | 96.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 mL
10 mL
50 mL
Packaging:
Glass bottle
Catalog number 162120025
also known as 16212
HSN 29159090
Price (INR)
8,501.00
Each
Quantity:
2.5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
8,501.00
Each
Chemical Identifiers
CAS381-73-7
IUPAC Name2,2-difluoroacetic acid
Molecular FormulaC2H2F2O2
InChI KeyPBWZKZYHONABLN-UHFFFAOYSA-N
SMILESOC(=O)C(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to light brown
Appearance (Form)Liquid
Infrared spectrumConforms
Titration with NaOH>=97.5 %
Refractive index1.3420 to 1.3450 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only