2-(Bromomethyl)naphthalene, 96%
2-(Bromomethyl)naphthalene, 96%
2-(Bromomethyl)naphthalene, 96%
Thermo Scientific Chemicals

2-(Bromomethyl)naphthalene, 96%

CAS: 939-26-4 | C11H9Br | 221.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 162560050
Price (USD)
53.65
Online exclusive
59.00
Save 5.35
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
53.65
Online exclusive
59.00
Save 5.35
Each
Chemical Identifiers
CAS939-26-4
IUPAC Name2-(bromomethyl)naphthalene
Molecular FormulaC11H9Br
InChI KeyRUHJZSZTSCSTCC-UHFFFAOYSA-N
SMILESBrCC1=CC=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow to light beige
Infrared spectrumConforms
GC>=95 %
Appearance (Form)Crystalline powder
Melting point52°C to 57°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only