delta-Valerolactone, 99%
delta-Valerolactone, 99%
delta-Valerolactone, 99%
Thermo Scientific Chemicals

delta-Valerolactone, 99%

CAS: 542-28-9 | C5H8O2 | 100.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number 162710010
Price (USD)
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS542-28-9
IUPAC Nameoxan-2-one
Molecular FormulaC5H8O2
InChI KeyOZJPLYNZGCXSJM-UHFFFAOYSA-N
SMILESO=C1CCCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Additional infoMay contain varying amounts of polymerized product
Additional infoMay contain varying amounts of polymerized product
Additional infoMay contain variable amounts of polymerized
Additional infoproduct
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only