4-Hydroxybenzaldehyde, 99%
4-Hydroxybenzaldehyde, 99%
4-Hydroxybenzaldehyde, 99%
Thermo Scientific Chemicals

4-Hydroxybenzaldehyde, 99%

CAS: 123-08-0 | C7H6O2 | 122.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number 162772500
Price (USD)
80.65
Online exclusive
89.80
Save 9.15
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
80.65
Online exclusive
89.80
Save 9.15
Each
Chemical Identifiers
CAS123-08-0
IUPAC Name4-hydroxybenzaldehyde
Molecular FormulaC7H6O2
InChI KeyRGHHSNMVTDWUBI-UHFFFAOYSA-N
SMILESOC1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.5 %
Melting point114°C to 118°C
Appearance (Color)Yellow to light brown
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only