Nunc™ MiniTrays with Nunclon™ Delta surface
Nunc™ MiniTrays with Nunclon™ Delta surface
Nunc™ MiniTrays with Nunclon™ Delta surface
Nunc™ MiniTrays with Nunclon™ Delta surface
Nunc™ MiniTrays with Nunclon™ Delta surface
Nunc™ MiniTrays with Nunclon™ Delta surface
Thermo Scientific™

Nunc™ MiniTrays with Nunclon™ Delta surface

Achieve excellent cell attachment and growth rates with Nunc MiniTrays with Nunclon Delta Surface.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWell Count
16311860
13652872
Catalog number 163118
Price (MXN)
-
Well Count:
60
Achieve excellent cell attachment and growth rates with Thermo Scientific™ Nunc™ MiniTrays with Nunclon™ Delta Surface.
  • Sterile polystyrene
  • Available with 60 or 72 conical wells
  • Large raised alphanumeric coordinates
  • Keyed lid for proper orientation

Specifications
Certifications/ComplianceCertified Nunclon surface
Closure IncludedYes
IncludesLid
Length (Metric)84.1 mm
No. per Case150
No. per Pack10
ShapeRectangular
TypeMini Tray
Volume (Metric) Working8 μL
Width (Metric)5.82 mm
Bottom ShapeConical
Capacity (Metric)8 L
Culture Area0.013 cm2
Height (Metric)11.2 mm
MaterialPolystyrene
SterilitySterile
Surface TreatmentNunclon Delta
Well Count60
Unit SizeCase of 150

Frequently asked questions (FAQs)

What reagents are used to coat your Nunclon Delta surface products?

The Nunclon Delta surface is a surface modification that enhances cell attachment and growth for adherent cell lines and is not coated with any chemical reagent.