2,4-Dichlorotoluene, 99%
2,4-Dichlorotoluene, 99%
2,4-Dichlorotoluene, 99%
Thermo Scientific Chemicals

2,4-Dichlorotoluene, 99%

CAS: 95-73-8 | C7H6Cl2 | 161.03 g/mol
Have Questions?
Catalog NumberQuantityPackaging
163282500250 mLGlass bottle
Catalog number 163282500
Price (USD)
43.65
Special offer
Online exclusive
Ends: 30-Jun-2026
51.10
Save 7.45 (15%)
Each
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
43.65
Special offer
Online exclusive
Ends: 30-Jun-2026
51.10
Save 7.45 (15%)
Each
Chemical Identifiers
CAS95-73-8
IUPAC Name2,4-dichloro-1-methylbenzene
Molecular FormulaC7H6Cl2
InChI KeyFUNUTBJJKQIVSY-UHFFFAOYSA-N
SMILESCC1=CC=C(Cl)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.5450 to 1.5470 (20°C, 589 nm)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only