Dimethylamine, 40 wt.% solution in water, Thermo Scientific Chemicals
Dimethylamine, 40 wt.% solution in water, Thermo Scientific Chemicals
Dimethylamine, 40 wt.% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethylamine, 40 wt.% solution in water, Thermo Scientific Chemicals

数量:
1 L
2.5 L
10 L
パッケージ:
すべて表示
Glass bottle
製品番号(カタログ番号) 163670025
価格(JPY)
-
数量:
2.5 L
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS124-40-3
IUPAC Namedimethylamine
Molecular FormulaC2H7N
InChI KeyROSDSFDQCJNGOL-UHFFFAOYSA-N
SMILESCNC
さらに表示
Appearance (Color)Clear colorless
Appearance (Form)Solution
Titration with HCl39.0 to 41.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only