Quinine, 98+%, anhydrous
Quinine, 98+%, anhydrous
Quinine, 98+%, anhydrous
Thermo Scientific Chemicals

Quinine, 98+%, anhydrous

CAS: 130-95-0 | C20H24N2O2 | 324.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 163702500
Price (USD)
-
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS130-95-0
IUPAC Name(R)-[(2S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol
Molecular FormulaC20H24N2O2
InChI KeyLOUPRKONTZGTKE-ZCXAXGJJNA-N
SMILES[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to almost white
Appearance (Form)Crystalline powder or needles
Infrared spectrumConforms
Melting point174°C to 179°C
Titration with HClO498.0 to 102.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only