Quinine sulfate dihydrate, 99+%
Quinine sulfate dihydrate, 99+%
Quinine sulfate dihydrate, 99+%
Thermo Scientific Chemicals

Quinine sulfate dihydrate, 99+%

CAS: 6119-70-6 | C40H54N4O10S | 782.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 163710100
Price (USD)
95.65
Online exclusive
106.00
Save 10.35
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
95.65
Online exclusive
106.00
Save 10.35
Each
Chemical Identifiers
CAS6119-70-6
IUPAC Namebis((R)-[(2S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol) sulfuric acid dihydrate
Molecular FormulaC40H54N4O10S
InChI KeyZHNFLHYOFXQIOW-OIGVVMIYNA-N
SMILESO.O.OS(O)(=O)=O.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C.[H][C@@]1(CN2CCC1C[C@@]2([H])[C@H](O)C1=C2C=C(OC)C=CC2=NC=C1)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals or needles
Infrared spectrumConforms
Titration with HClO499.0 to 101.0 % (on anhydrous basis)
Water3.0 to 6.0 % (K.F.)
Appearance (Color)White to almost white
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only