Ethyl 4-chloro-2-methylthio-5-pyrimidinecarboxylate, 98%
Ethyl 4-chloro-2-methylthio-5-pyrimidinecarboxylate, 98%
Ethyl 4-chloro-2-methylthio-5-pyrimidinecarboxylate, 98%
Thermo Scientific Chemicals

Ethyl 4-chloro-2-methylthio-5-pyrimidinecarboxylate, 98%

CAS: 5909-24-0 | C8H9ClN2O2S | 232.68 g/mol
Have Questions?
Catalog NumberQuantityPackaging
163751000100 gGlass bottle
Catalog number 163751000
Price (USD)
523.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
523.00
Each
Chemical Identifiers
CAS5909-24-0
IUPAC Nameethyl 4-chloro-2-(methylsulfanyl)pyrimidine-5-carboxylate
Molecular FormulaC8H9ClN2O2S
InChI KeySNNHLSHDDGJVDM-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CN=C(SC)N=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Melting point59°C to 64°C
Appearance (Color)White to yellow
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only