5-Methylsalicylic acid, 98%
5-Methylsalicylic acid, 98%
5-Methylsalicylic acid, 98%
Thermo Scientific Chemicals

5-Methylsalicylic acid, 98%

CAS: 89-56-5 | C8H8O3 | 152.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
16386010010 gGlass bottle
Catalog number 163860100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS89-56-5
IUPAC Name2-hydroxy-5-methylbenzoic acid
Molecular FormulaC8H8O3
InChI KeyDLGBEGBHXSAQOC-UHFFFAOYSA-N
SMILESCC1=CC=C(O)C(=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=97.5 %
Melting point151°C to 155°C
Appearance (Color)White to beige
Infrared spectrumConforms
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only