Bathophenanthrolinedisulfonic acid, disodium salt hydrate, 97%
Bathophenanthrolinedisulfonic acid, disodium salt hydrate, 97%
Bathophenanthrolinedisulfonic acid, disodium salt hydrate, 97%
Thermo Scientific Chemicals

Bathophenanthrolinedisulfonic acid, disodium salt hydrate, 97%

CAS: 52746-49-3 | C24H14N2Na2O6S2 | 536.48 g/mol
Quantity:
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 164050050
Price (JPY)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS52746-49-3
IUPAC Namedisodium 4-[7-(4-sulfonatophenyl)-1,10-phenanthrolin-4-yl]benzene-1-sulfonate
Molecular FormulaC24H14N2Na2O6S2
InChI KeyPCNDSIWXTYFWIA-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)C1=CC=NC2=C1C=CC1=C2N=CC=C1C1=CC=C(C=C1)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige to pink to orange
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=96.0 %
Water2 to 12 % (K.F.)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only