2,4-Dichlorobenzaldehyde, 98%
2,4-Dichlorobenzaldehyde, 98%
2,4-Dichlorobenzaldehyde, 98%
Thermo Scientific Chemicals

2,4-Dichlorobenzaldehyde, 98%

CAS: 874-42-0 | C7H4Cl2O | 175.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Plastic bottle
Catalog number 164141000
Price (USD)
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS874-42-0
IUPAC Name2,4-dichlorobenzaldehyde
Molecular FormulaC7H4Cl2O
InChI KeyYSFBEAASFUWWHU-UHFFFAOYSA-N
SMILESClC1=CC=C(C=O)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or chunks
Infrared spectrumConforms
Appearance (Color)Light yellow to beige
Melting point68°C to 73°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only