2-Bromobutyric acid, 99%
2-Bromobutyric acid, 99%
2-Bromobutyric acid, 99%
Thermo Scientific Chemicals

2-Bromobutyric acid, 99%

CAS: 80-58-0 | C4H7BrO2 | 167.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
View all
Glass bottle
Catalog number 164510010
Price (EUR)
346,00
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
346,00
Each
Chemical Identifiers
CAS80-58-0
IUPAC Name2-bromobutanoic acid
Molecular FormulaC4H7BrO2
InChI KeyYAQLSKVCTLCIIE-UHFFFAOYNA-N
SMILESCCC(Br)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow
GC>=98.5 %
Refractive index1.4715 to 1.4755 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only