trans-4-Phenyl-3-buten-2-one, 99%
trans-4-Phenyl-3-buten-2-one, 99%
trans-4-Phenyl-3-buten-2-one, 99%
Thermo Scientific Chemicals

trans-4-Phenyl-3-buten-2-one, 99%

CAS: 1896-62-4 | C10H10O | 146.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 164520500
Price (USD)
40.65
Special offer
Online exclusive
Ends: 28-Dec-2026
47.10
Save 6.45
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
40.65
Special offer
Online exclusive
Ends: 28-Dec-2026
47.10
Save 6.45
Each
Chemical Identifiers
CAS1896-62-4
IUPAC Name(3E)-4-phenylbut-3-en-2-one
Molecular FormulaC10H10O
InChI KeyBWHOZHOGCMHOBV-BQYQJAHWSA-N
SMILESCC(=O)\C=C\C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point38.5°C to 41°C
Appearance (Form)Powder or crystals or solid
Appearance (Color)White to yellow
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only