PepSwift™ Monolithic Capillary HPLC Columns
Thermo Scientific™

PepSwift™ Monolithic Capillary HPLC Columns

Use for fast, high-resolution LC/MS separations in bottom-up protein identification, biomarker discovery, and systems biology applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)
164542250 mm0.2 mm
16458450 mm0.1 mm
Catalog number 164542
Price (USD)
1,607.55
Each
Length (Metric):
250 mm
Diameter (Metric):
0.2 mm
Price (USD)
1,607.55
Each
For fast, high-resolution LC/MS separations in bottom-up protein identification, biomarker discovery, and systems biology applications, use Thermo Scientific™ PepSwift™ Monolithic Capillary LC Columns. Based on a polystyrene divinylbenzene copolymer, the monolithic structure offers a high-quality alternative to traditional microparticulate sorbents, providing important advantages to the chromatographic separation. High-sensitivity proteomics and biotech applications are easily performed using these columns.

Benefit from:

  • High-speed peptide and protein separations (<15 min)
  • High sensitivity for LC/MS
  • High column-to-column reproducibility
  • Wide range of column I.D. and lengths available
  • Superior lifetime
  • Easy column installation using Thermo Scientific™ Dionex™ nanoViper™ Fingertight Fittings
  • Precolumns available to preconcentrate and desalt samples

You can also use Thermo Scientific™ PepSwift™ Trap Columns for preconcentration and desalting of samples consisting of peptides and proteins without negatively impacting the chromatographic performance or recovery of the compounds. Change the selectivity of the separation or improve the trapping efficiency by employing various ion-pairing agents in the loading solvent and/or mobile phases. The trap columns are used with Thermo Scientific™ PepSwift™ and ProSwift™ Monolithic Columns, which are for fast, high-resolution LC/MS separations in protein identification, biomarker discovery, and systems biology applications.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatCapillary Column, NanoViper
Column TypeReversed Phase
Max. Pressure4350 psi (300 bar)
Packing MaterialMonolith Polymer
Product TypeAnalytical Column
Stationary PhasePSDVB (Polystyrene Divinylbenzene)
Temperature80°C
Diameter (Metric)0.2 mm
Length (Metric)250 mm
MaterialPolystyrene, Divinylbenzene
Particle SizeMonolith
PhaseReversed Phase
Pore SizeMonolith
Product LinenanoLC
pH1 to 10
Unit SizeEach