2-Ethyl-2-(hydroxymethyl)-1,3-propanediol, 98%
2-Ethyl-2-(hydroxymethyl)-1,3-propanediol, 98%
2-Ethyl-2-(hydroxymethyl)-1,3-propanediol, 98%
Thermo Scientific Chemicals

2-Ethyl-2-(hydroxymethyl)-1,3-propanediol, 98%

CAS: 77-99-6 | C6H14O3 | 134.18 g/mol
数量:
500 g
1 kg
2.5 kg
10 kg
パッケージ:
すべて表示
Plastic bottle
製品番号(カタログ番号) 164655000
価格(JPY)
-
数量:
500 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS77-99-6
IUPAC Name2-ethyl-2-(hydroxymethyl)propane-1,3-diol
Molecular FormulaC6H14O3
InChI KeyZJCCRDAZUWHFQH-UHFFFAOYSA-N
SMILESCCC(CO)(CO)CO
さらに表示
Appearance (Form)Flakes
Melting point>=57°C
Ash=<0.0015 %
Acidity=<0.002 % (as Formic acid)
Water=<0.1 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only