4-Methylanisole, 99%, Thermo Scientific Chemicals
4-Methylanisole, 99%, Thermo Scientific Chemicals
4-Methylanisole, 99%, Thermo Scientific Chemicals

4-Methylanisole, 99%, Thermo Scientific Chemicals

CAS: 104-93-8 | C8H10O | 122.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number 164662500
Price (USD)
53.80
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
53.80
Each
Chemical Identifiers
CAS104-93-8
IUPAC Name1-methoxy-4-methylbenzene
Molecular FormulaC8H10O
InChI KeyCHLICZRVGGXEOD-UHFFFAOYSA-N
SMILESCOC1=CC=C(C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5100 to 1.5130 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only