Nunc™ 384 Well ShallowWell Standard Height Black and White Plates - FAQs
Nunc™ 384 Well ShallowWell Standard Height Black and White Plates - FAQs
View additional product information for Nunc™ 384 Well ShallowWell Standard Height Black and White Plates - FAQs (164702)
2 product FAQs found
Do you offer Nunc Microplate Lids for the Nunc Black or White 384-Well Microplates?
We offer Nunc Microplate Lids: Cat. No. 448143, that fit both the 384-well microplates and the 1,536-well microplates.
Why do you offer Nunc 384-Well Black or White Microplates when the cells cannot be observed using an inverted light microscope?
We offer Nunc Black or White 384-Well microplates that are intended for use in conjunction with 384-well microplate luminometers and fluorometers that have excitation and detection systems located above the plate. Due to the opaque quality of these microplates, all detection systems must rely on either luminescent or fluorescent reagents, such as viability stains that report cell activity and membrane integrity.