Thiobenzamide, 95%
Thiobenzamide, 95%
Thiobenzamide, 95%
Thermo Scientific Chemicals

Thiobenzamide, 95%

CAS: 2227-79-4 | C7H7NS | 137.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 164730100
Price (USD)
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS2227-79-4
IUPAC Namebenzenecarbothioamide
Molecular FormulaC7H7NS
InChI KeyQIOZLISABUUKJY-UHFFFAOYSA-N
SMILESNC(=S)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder and/or chunks
HPLC>=94 %
Appearance (Color)Yellow-green
Infrared spectrumConforms
Melting point114°C to 118°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only