2-Methoxynaphthalene, 98%
2-Methoxynaphthalene, 98%
2-Methoxynaphthalene, 98%
Thermo Scientific Chemicals

2-Methoxynaphthalene, 98%

CAS: 93-04-9 | C11H10O | 158.2 g/mol
数量:
5 g
500 g
パッケージ:
すべて表示
Glass bottle
製品番号(カタログ番号) 164750050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS93-04-9
IUPAC Name2-methoxynaphthalene
Molecular FormulaC11H10O
InChI KeyLUZDYPLAQQGJEA-UHFFFAOYSA-N
SMILESCOC1=CC=C2C=CC=CC2=C1
さらに表示
Appearance (Color)White to yellow-brown
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Crystalline solid
Melting point>=71.5°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only