2-Methoxynaphthalene, 98%
2-Methoxynaphthalene, 98%
2-Methoxynaphthalene, 98%
Thermo Scientific Chemicals

2-Methoxynaphthalene, 98%

CAS: 93-04-9 | C11H10O | 158.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 g
Packaging:
View all
Plastic bottle
Catalog number 164755000
Price (USD)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS93-04-9
IUPAC Name2-methoxynaphthalene
Molecular FormulaC11H10O
InChI KeyLUZDYPLAQQGJEA-UHFFFAOYSA-N
SMILESCOC1=CC=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow-brown
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Crystalline solid
Melting point>=71.5°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only