UltraPure™ Agarose, 100 g - FAQs
UltraPure™ Agarose, 100 g - FAQs
View additional product information for UltraPure™ Agarose - FAQs (16500500, 16500100)
1 product FAQs found
I want to pour my own gels. Which agarose should I use?
Our UltraPure Agarose is standard melting-point agarose designed for routine separation and analysis of DNA and RNA fragments in the 500-23,000 nt range. UltraPure Agarose 1000 is a specialized agarose that provides higher resolution of PCR fragments and other short DNA fragments. We also offer an UltraPure Low Melting Point Agarose, which is ideal for resolving DNA fragments from 10 to 1,000 bp with a low melting temperature of 65°C or less.