Omnitray 128x86mm Single Well Plate, TC Surface, Pack of 10 - FAQs
Omnitray 128x86mm Single Well Plate, TC Surface, Pack of 10 - FAQs
View additional product information for Nunc™ OmniTray™ Single-Well Plate - FAQs (140156, 165218, 242811, 264728)
2 product FAQs found
What is the difference between the Nunclon Delta surface and a tissue culture (TC) surface from a competitor?
Our Nunclon Delta surface for adherent cells is a standard tissue culture (TC) surface modification that makes the polystyrene surface more hydrophilic. This facilitates maximum adhesion for a broad range of cell types.
The Nunclon Delta surface is Thermo Scientific Nunclon Delta-certified, which is a Thermo Fisher proprietary certification that has a quality standard to help ensure consistency. Every lot of Thermo Scientific Nunclon Delta-certified cell cultureware is tested with Gibco media for:
- Less than 10% growth variation in L929, Hel299 and primary chick embryo cells
- Less than 15% variation colony formation of V79-4 cells
More information and all applicable products can be found in the "Product selection guide for every step of your cell culture workflow" brochure .
When screening DNA libraries, how much agar should I add to the Nunc OmniTray Single-Well Plate?
Our Nunc OmniTray Single-Well Plate can hold 35 mL of growth media. We recommend adding 1.5% agar to the growth media.