Accucore™ 150-C4 HPLC Columns
Thermo Scientific™

Accucore™ 150-C4 HPLC Columns

Use this specially designed solid core column with a C4 functionality for biomolecules that bind too tightly to C8 or C18 columns.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)
16526-103030100 mm3 mm
16526-03213030 mm2.1 mm
16526-05213050 mm2.1 mm
16526-102130100 mm2.1 mm
16526-104630100 mm4.6 mm
16526-152130150 mm2.1 mm
16526-154630150 mm4.6 mm
16526-157569150 mm0.075 mm
16526-01210510 mm2.1 mm
16526-01300510 mm3 mm
16526-01400510 mm4 mm
Catalog number 16526-103030
Price (USD)
1,127.14
Each
Length (Metric):
100 mm
Diameter (Metric):
3 mm
Price (USD)
1,127.14
Each

Offer lower hydrophobic retention, ideal for optimal retention of proteins and larger peptides with Thermo Scientific™ Accucore™ 150-C4 LC Columns. Based on Core Enhanced Technology, Thermo Scientific Accucore columns provide fast, high-resolution biomolecule separations without the elevated backpressures required by sub-2μm particles.

  • 150Å pore size solid core particles for fast biomoleucule separations
  • Superb resolution at low backpressures
  • Exceptionally rugged columns
  • Significantly lower hydrophobic retention than C18
  • Ideal for optimal retention of proteins and larger peptides

Ordering Information:

Other custom column dimensions are available. Please call your local Customer Service for more information.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
EndcappedYes
For Use WithHPLC, UHPLC
Max. Pressure14500 psi (1000 bar)
Packing MaterialUltrapure Silica
Particle ShapeSolid Core
QuantityEach
Stationary PhaseC4
Surface Area80 m²/g
Temperature60°C
USP TypeL26
Carbon Load2%
Diameter (Metric)3 mm
Length (Metric)100 mm
Particle Size2.6 μm
PhaseReversed Phase
Pore Size150 Å
Product LineAccucore
pH2 to 9
Unit SizeEach