Thiazolidine, 98%
Thiazolidine, 98%
Thiazolidine, 98%
Thermo Scientific Chemicals

Thiazolidine, 98%

CAS: 504-78-9 | C3H8NS | 90.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 165310250
Price (EUR)
355,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
355,00
Each
Chemical Identifiers
CAS504-78-9
IUPAC Name1,3-thiazolidin-3-ium
Molecular FormulaC3H8NS
InChI KeyOGYGFUAIIOPWQD-UHFFFAOYSA-O
SMILESC1CSC[NH2+]1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.5525 to 1.5545 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only