Hexahydro-4-methylphthalic anhydride, 98%, mixture of cis and trans
Hexahydro-4-methylphthalic anhydride, 98%, mixture of cis and trans
Hexahydro-4-methylphthalic anhydride, 98%, mixture of cis and trans
Thermo Scientific Chemicals

Hexahydro-4-methylphthalic anhydride, 98%, mixture of cis and trans

CAS: 19438-60-9 | C9H12O3 | 168.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 165390025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS19438-60-9
IUPAC Name5-methyl-octahydro-2-benzofuran-1,3-dione
Molecular FormulaC9H12O3
InChI KeyFKBMTBAXDISZGN-UHFFFAOYNA-N
SMILESCC1CCC2C(C1)C(=O)OC2=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4764 to 1.4784 (20°C, 589 nm)
GC>=97.5 %
Infrared spectrumConforms
Color scale=<100 APHA
Appearance (Form)Clear liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only