Methyl disulfide, 99%
Methyl disulfide, 99%
Methyl disulfide, 99%
Thermo Scientific Chemicals

Methyl disulfide, 99%

CAS: 624-92-0 | C2H6S2 | 94.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number 165590010
Price (USD)
80.65
Special offer
Online exclusive
Ends: 28-Dec-2026
94.80
Save 14.15
Each
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
80.65
Special offer
Online exclusive
Ends: 28-Dec-2026
94.80
Save 14.15
Each
Chemical Identifiers
CAS624-92-0
IUPAC Name(methyldisulfanyl)methane
Molecular FormulaC2H6S2
InChI KeyWQOXQRCZOLPYPM-UHFFFAOYSA-N
SMILESCSSC
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5235 to 1.5265 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only