5alpha-Cholestane, 98+%
5alpha-Cholestane, 98+%
5alpha-Cholestane, 98+%
Thermo Scientific Chemicals

5alpha-Cholestane, 98+%

CAS: 481-21-0 | C27H48 | 372.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Catalog number 165600010
Price (USD)
161.65
Special offer
Online exclusive
Ends: 28-Dec-2026
190.00
Save 28.35
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
161.65
Special offer
Online exclusive
Ends: 28-Dec-2026
190.00
Save 28.35
Each
Chemical Identifiers
CAS481-21-0
IUPAC Name9a,11a-dimethyl-1-(6-methylheptan-2-yl)-hexadecahydro-1H-cyclopenta[a]phenanthrene
Molecular FormulaC27H48
InChI KeyXIIAYQZJNBULGD-UHFFFAOYNA-N
SMILESCC(C)CCCC(C)C1CCC2C3CCC4CCCCC4(C)C3CCC12C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point78°C to 81°C
Specific optical rotation+23° to +30° (20°C, 589 nm) (c=2, CHCl3)
Appearance (Color)White
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only