This product ships via ground from US northeast.
DL-1,4-Dithiothreitol, 99%, for biochemistry
DL-1,4-Dithiothreitol, 99%, for biochemistry
DL-1,4-Dithiothreitol, 99%, for biochemistry
Thermo Scientific Chemicals

DL-1,4-Dithiothreitol, 99%, for biochemistry

1,4-Dithiothreitol, CAS # 3483-12-3, is a reducing agent commonly used to reduce disulphide bonds in protein studies, such as western blot SDS-page electrophoresis. | CAS: 3483-12-03 | C4H10O2S2 | 154.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 165681000
Price (USD)
1,425.65
Online Exclusive
1,584.00
Save 158.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
1,425.65
Online Exclusive
1,584.00
Save 158.35 (10%)
Each
Chemical Identifiers
CAS3483-12-3
IUPAC Name(2R,3R)-1,4-disulfanylbutane-2,3-diol
Molecular FormulaC4H10O2S2
InChI KeyVHJLVAABSRFDPM-IMJSIDKUSA-N
SMILESO[C@@H](CS)[C@@H](O)CS
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point38°C to 43°C
Titration Iodimetric98.5 to 101.5 %
UV(1 M in water)
UV transmittance>=99 % (c=5, 660 nm, water) 1 cm cell
UVat 500 nm A: =<0.035
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • 1,4-Dithiothreitol (DTT or Cleland's reagent) reduces disulfide bonds in proteins, leading to loss of native conformation
  • DTT is not stable in solution and should be prepared in small aliquots prior to use; the use of EDTA in the solution can increase the half-life of DTT
  • Typically used at a final concentration of 0.1M for protein stabilization; recommended to create a 1M stock

Applications

  • Protects thiol groups from oxidation and acts as a chemical chelator in solution; maintains proteins in the denatured state
  • Common applications include SDS-page electrophoresis and protein isolation
RUO – Research Use Only