Tetra Electrophoresis Core Set
Invitrogen™
Tetra Electrophoresis Core Set
The Tetra Electrophoresis Core Set includes both the Tetra Electrophoresis primary and companion cores, enabling you to run up to four Invitrogen protein gels in Bio-Rad's Mini-PROTEAN™ Tetra Cell tank. Achieve high-performance separations and fast run times with this comprehensive set.
Have Questions?
Catalog NumberQuantity
16580401 Set
Catalog number 1658040
Price (USD)
376.00
Each
Quantity:
1 Set
Request bulk or custom format
Price (USD)
376.00
Each
The Invitrogen Tetra Electrophoresis Core Set allows users to run Invitrogen precast protein gels without the need to purchase a new tank or adopt a new workflow. Designed for seamless integration with Bio-Rad's Mini-PROTEAN Tetra Cell, it provides an easy and straightforward method of adopting Invitrogen precast mini gels for protein analysis applications such as western blotting and SDS-PAGE. The Core Set includes both the Tetra Electrophoresis primary and companion cores. Its intuitive design ensures ease of use, making it accessible for both new and experienced users.
  • Swap and go—designed to work seamlessly with Bio-Rad's Mini-PROTEAN Tetra Cell, allowing for a smooth transition between gel types
  • Achieve high performance—compatible with Invitrogen precast mini protein gels for high-quality results backed by our protein gel performance guarantee
  • Run up to four gels simultaneously—capable of running up to four gels per run to increase sample throughput
  • Access multiple gel chemistries—compatible with all Invitrogen mini gels for diverse applications
The Tetra Electrophoresis cores stand out due to their compatibility with all Invitrogen mini gels, allowing users to swap and run gels effortlessly while achieving high-performance results. When using both cores, up to four gels can be run simultaneously, which significantly increases throughput and efficiency. Additionally, the self-aligning cassettes and leak-proof design ensure stable and reliable electrophoresis runs, making experiment setup easy and fast.
The Tetra Electrophoresis Primary Core comes with the Tetra Buffer Dam (Cat. No. 1658039) for use when running only one gel in the core. To run three or four gels at once, the Tetra Electrophoresis Companion Core (Cat. No. 1658038) is used alongside the Primary Core. This combination allows for the simultaneous running of multiple gels, further enhancing throughput and efficiency in your lab. The Companion Core works seamlessly with the Primary Core, providing a flexible solution for varying experimental needs.
This cost-effective solution eliminates the need for new electrophoresis equipment, saving costs while enhancing your lab's capabilities. The Tetra Electrophoresis Core Set is straightforward to use: simply swap the cores in your Bio-Rad Mini-PROTEAN Tetra system for the Invitrogen Tetra cores and use Invitrogen mini protein gels following their recommended protocols. Invitrogen precast protein gels offer excellent protein integrity with neutral pH, a wide range of molecular weight separation, fast run times, and a long shelf life, making them ideal for high-performance and reproducible results in your experiments.
For Research Use Only.
Specifications
Mode of SeparationMolecular Weight
Quantity1 Set
For Use With (Equipment)Bio-Rad Mini-PROTEAN Tetra Cell
TypeElectrophoresis Core
Unit SizeEach