Glycidyl methacrylate, 97%, stabilized
Glycidyl methacrylate, 97%, stabilized
Glycidyl methacrylate, 97%, stabilized
Thermo Scientific Chemicals

Glycidyl methacrylate, 97%, stabilized

CAS: 106-91-2 | C7H10O3 | 142.15 g/mol
数量:
100 g
500 g
2.5 kg
パッケージ:
Glass bottle
製品番号(カタログ番号) 165895000
価格(JPY)
-
数量:
500 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS106-91-2
IUPAC Name(oxiran-2-yl)methyl 2-methylprop-2-enoate
Molecular FormulaC7H10O3
InChI KeyVOZRXNHHFUQHIL-UHFFFAOYNA-N
SMILESCC(=C)C(=O)OCC1CO1
さらに表示
Infrared spectrumConforms
Water=<0.1 % (K.F.)
Refractive index1.4490 to 1.4510 (20°C, 589 nm)
Appearance (Form)Clear liquid
Color scale=<30 APHA
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only