Thiazole, 99%
Thiazole, 99%
Thiazole, 99%
Thermo Scientific Chemicals

Thiazole, 99%

CAS: 288-47-1 | C3H3NS | 85.12 g/mol
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 166130250
Price (JPY)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS288-47-1
IUPAC Name1,3-thiazole
Molecular FormulaC3H3NS
InChI KeyFZWLAAWBMGSTSO-UHFFFAOYSA-N
SMILESS1C=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.5365 to 1.5395 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only