Trifluoroacetic acid-d, for NMR, 99 atom % D
Trifluoroacetic acid-d, for NMR, 99 atom % D
Trifluoroacetic acid-d, for NMR, 99 atom % D
Thermo Scientific Chemicals

Trifluoroacetic acid-d, for NMR, 99 atom % D

CAS: 599-00-8 | C2HF3O2 | 115.029 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
10 mL
25 mL
100 mL
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 166400100
Price (USD)
-
Quantity:
10 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS599-00-8
IUPAC Nametrifluoroethan(²H)oic acid
Molecular FormulaC2HF3O2
InChI KeyDTQVDTLACAAQTR-DYCDLGHISA-N
SMILES[2H]OC(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
D-Enrichment>=98.5 %
Water=<0.1 % (Coulometric)
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only