2,3,4-Trimethoxybenzaldehyde, 99%
2,3,4-Trimethoxybenzaldehyde, 99%
2,3,4-Trimethoxybenzaldehyde, 99%
Thermo Scientific Chemicals

2,3,4-Trimethoxybenzaldehyde, 99%

CAS: 2103-57-3 | C10H12O4 | 196.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 166450100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2103-57-3
IUPAC Name2,3,4-trimethoxybenzaldehyde
Molecular FormulaC10H12O4
InChI KeyUCTUXUGXIFRVGX-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=O)C(OC)=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point>=36.5°C
GC>=98.5 %
Appearance (Color)White to light yellow
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only