Isocarbostyril, 98%
Isocarbostyril, 98%
Isocarbostyril, 98%
Thermo Scientific Chemicals

Isocarbostyril, 98%

CAS: 491-30-5 | C9H7NO | 145.16 g/mol
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 166460010
Price (JPY)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS491-30-5
IUPAC Name1,2-dihydroisoquinolin-1-one
Molecular FormulaC9H7NO
InChI KeyVDBNYAPERZTOOF-UHFFFAOYSA-N
SMILESO=C1NC=CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Appearance (Form)Powder
Infrared spectrumConforms
Melting point210°C to 215°C
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only