BioAssay Dish 500cm2, 245x245mm, TC Surface, Pack of 4 - FAQs
BioAssay Dish 500cm2, 245x245mm, TC Surface, Pack of 4 - FAQs
View additional product information for Nunc™ Square BioAssay Dishes - FAQs (166508, 240835, 240845)
2 product FAQs found
What is the difference between the Nunclon Delta surface and a tissue culture (TC) surface from a competitor?
Our Nunclon Delta surface for adherent cells is a standard tissue culture (TC) surface modification that makes the polystyrene surface more hydrophilic. This facilitates maximum adhesion for a broad range of cell types.
The Nunclon Delta surface is Thermo Scientific Nunclon Delta-certified, which is a Thermo Fisher proprietary certification that has a quality standard to help ensure consistency. Every lot of Thermo Scientific Nunclon Delta-certified cell cultureware is tested with Gibco media for:
- Less than 10% growth variation in L929, Hel299 and primary chick embryo cells
- Less than 15% variation colony formation of V79-4 cells
More information and all applicable products can be found in the "Product selection guide for every step of your cell culture workflow" brochure .
Do you offer a Nunc square petri dish with a grid on the bottom?
We offer Nunc Square BioAssay Dishes, but this product does not have a grid.