1-Bromooctane, 99%
1-Bromooctane, 99%
1-Bromooctane, 99%
Thermo Scientific Chemicals

1-Bromooctane, 99%

CAS: 111-83-1 | C8H17Br | 193.13 g/mol
数量:
500 mL
2.5 L
パッケージ:
Glass bottle
製品番号(カタログ番号) 166815000
価格(JPY)
-
数量:
500 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS111-83-1
IUPAC Name1-bromooctane
Molecular FormulaC8H17Br
InChI KeyVMKOFRJSULQZRM-UHFFFAOYSA-N
SMILESCCCCCCCCBr
さらに表示
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4508 to 1.4528 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow-brown
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only