Lauric acid, 99%
Lauric acid, 99%
Lauric acid, 99%
Thermo Scientific Chemicals

Lauric acid, 99%

CAS: 143-07-7 | C12H24O2 | 200.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 167280010
Price (USD)
146.65
Special offer
Online exclusive
Ends: 28-Dec-2026
172.00
Save 25.35
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
146.65
Special offer
Online exclusive
Ends: 28-Dec-2026
172.00
Save 25.35
Each
Chemical Identifiers
CAS143-07-7
IUPAC Namedodecanoic acid
Molecular FormulaC12H24O2
InChI KeyPOULHZVOKOAJMA-UHFFFAOYSA-N
SMILESCCCCCCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or flakes
GC>=98.5 %
Infrared spectrumConforms
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only