5-Bromo-2-anisaldehyde, 99%
5-Bromo-2-anisaldehyde, 99%
5-Bromo-2-anisaldehyde, 99%
Thermo Scientific Chemicals

5-Bromo-2-anisaldehyde, 99%

CAS: 25016-01-7 | C8H7BrO2 | 215.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 167520050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS25016-01-7
IUPAC Name5-bromo-2-methoxybenzaldehyde
Molecular FormulaC8H7BrO2
InChI KeyIJIBRSFAXRFPPN-UHFFFAOYSA-N
SMILESCOC1=CC=C(Br)C=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Appearance (Color)Off-white to yellow to beige
Melting point115°C to 119°C
GC>=98.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only