Tris(hydroxymethyl)aminomethane, 99.8%, for analysis, biochemical grade
Tris(hydroxymethyl)aminomethane, 99.8%, for analysis, biochemical grade
Tris(hydroxymethyl)aminomethane, 99.8%, for analysis, biochemical grade
Thermo Scientific Chemicals

Tris(hydroxymethyl)aminomethane, 99.8%, for analysis, biochemical grade

CAS: 77-86-1 | C4H11NO3 | 121.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
5 kg
Packaging:
View all
Plastic drum
Catalog number 167620051
Price (USD)
377.65
Special offer
Online exclusive
Ends: 28-Dec-2026
444.00
Save 66.35 (15%)
Each
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)
377.65
Special offer
Online exclusive
Ends: 28-Dec-2026
444.00
Save 66.35 (15%)
Each
Chemical Identifiers
CAS77-86-1
IUPAC Name2-amino-2-(hydroxymethyl)propane-1,3-diol
Molecular FormulaC4H11NO3
InChI KeyLENZDBCJOHFCAS-UHFFFAOYSA-N
SMILESNC(CO)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
UV(10 % in water) 1 cm cell
UVat 260 nm A: =<0.1
Iron (Fe)=<1 ppm
Appearance (Form)Crystalline powder or crystals and/or chunks
Sulfated ash=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only