Acetonitrile, 99+%, for spectroscopy
Acetonitrile, 99+%, for spectroscopy
Acetonitrile, 99+%, for spectroscopy
Acros Organics

Acetonitrile, 99+%, for spectroscopy

CAS: 75-05-8 | C2H3N | 41.053 g/mol
Have Questions?
Catalog NumberQuantityPackaging
167652500
also known as 16765
250 mLGlass Bottle
Catalog number 167652500
also known as 16765
HSN 29269000
Price (INR)
7,175.00
Each
Quantity:
250 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
7,175.00
Each
Chemical Identifiers
CAS75-05-8
IUPAC Nameacetonitrile
Molecular FormulaC2H3N
InChI KeyWEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILESCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
GC>=99.0 %
UVat 193 nm A: =<1
UVat 210 nm A: =<0.3
UVat 235 nm A: =<0.02
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
RUO – Research Use Only