Benzonitrile, 99+%, for spectroscopy
Benzonitrile, 99+%, for spectroscopy
Benzonitrile, 99+%, for spectroscopy
Thermo Scientific Chemicals

Benzonitrile, 99+%, for spectroscopy

CAS: 100-47-0 | C7H5N | 103.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number 167670500
Price (USD)
35.65
Special offer
Online exclusive
Ends: 28-Dec-2026
41.80
Save 6.15
Each
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
35.65
Special offer
Online exclusive
Ends: 28-Dec-2026
41.80
Save 6.15
Each
Chemical Identifiers
CAS100-47-0
IUPAC Namebenzonitrile
Molecular FormulaC7H5N
InChI KeyJFDZBHWFFUWGJE-UHFFFAOYSA-N
SMILESN#CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
UVat 310 nm A: =<0.15
UVat 360 nm A: =<0.01
Water=<0.03 %
Appearance (Form)Clear liquid
UVat 300 nm A: =<1
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
RUO – Research Use Only