1-Butanol, 99+%, for spectroscopy
1-Butanol, 99+%, for spectroscopy
1-Butanol, 99+%, for spectroscopy
Thermo Scientific Chemicals

1-Butanol, 99+%, for spectroscopy

CAS: 71-36-3 | C4H10O | 74.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass Bottle
Catalog number 167691000
Price (USD)
37.65
Special offer
Online exclusive
Ends: 28-Dec-2026
44.70
Save 7.05
Each
Quantity:
100 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
37.65
Special offer
Online exclusive
Ends: 28-Dec-2026
44.70
Save 7.05
Each
Chemical Identifiers
CAS71-36-3
IUPAC Namebutan-1-ol
Molecular FormulaC4H10O
InChI KeyLRHPLDYGYMQRHN-UHFFFAOYSA-N
SMILESCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
GC>=99.0 %
UVat 210 nm A: =<0.602
UVat 220 nm A: =<0.400
UVat 238 nm A: =<0.097
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
RUO – Research Use Only