1,4-Dioxane, 99+%, for spectroscopy
1,4-Dioxane, 99+%, for spectroscopy
1,4-Dioxane, 99+%, for spectroscopy
Thermo Scientific Chemicals

1,4-Dioxane, 99+%, for spectroscopy

CAS: 123-91-1 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number 167800025
Price (USD)
-
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS123-91-1
IUPAC Name1,4-dioxane
Molecular FormulaC4H8O2
InChI KeyRYHBNJHYFVUHQT-UHFFFAOYSA-N
SMILESC1COCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
GC>=99.0 %
Water=<0.03 %
Appearance (Form)Clear liquid
UVat 225 nm A: =<0.5
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only