Ethyl acetate, 99.5+%, for spectroscopy
Ethyl acetate, 99.5+%, for spectroscopy
Ethyl acetate, 99.5+%, for spectroscopy
Thermo Scientific Chemicals

Ethyl acetate, 99.5+%, for spectroscopy

CAS: 141-78-6 | C4H8O2 | 88.106 g/mol
数量:
500 mL
1 L
2.5 L
パッケージ:
Glass Bottle
製品番号(カタログ番号) 167810025
価格(JPY)
-
数量:
2.5 L
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS141-78-6
IUPAC Nameethyl acetate
Molecular FormulaC4H8O2
InChI KeyXEKOWRVHYACXOJ-UHFFFAOYSA-N
SMILESCCOC(C)=O
さらに表示
Residue after evaporation=<0.0003 %
Water=<0.05 %
UVat 255 nm A: =<1
UVat 275 nm A: =<0.05
Appearance (Form)Clear liquid
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
RUO – Research Use Only