2-Piperidinemethanol, 97%
2-Piperidinemethanol, 97%
2-Piperidinemethanol, 97%
Thermo Scientific Chemicals

2-Piperidinemethanol, 97%

CAS: 3433-37-2 | C6H13NO | 115.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 168030050
Price (EUR)
45,10
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
45,10
Each
Chemical Identifiers
CAS3433-37-2
IUPAC Name(2R)-2-(hydroxymethyl)piperidin-1-ium
Molecular FormulaC6H14NO
InChI KeyPRAYXGYYVXRDDW-ZCFIWIBFSA-O
SMILESOC[C@H]1CCCC[NH2+]1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solid
Appearance (Color)White to light orange
GC>=96.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only