3-Piperidinemethanol, 98%
3-Piperidinemethanol, 98%
3-Piperidinemethanol, 98%
Thermo Scientific Chemicals

3-Piperidinemethanol, 98%

CAS: 4606-65-9 | C6H13NO | 115.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 168040050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4606-65-9
IUPAC Name(3R)-3-(hydroxymethyl)piperidin-1-ium
Molecular FormulaC6H14NO
InChI KeyVUNPWIPIOOMCPT-ZCFIWIBFSA-O
SMILESOC[C@@H]1CCC[NH2+]C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)White to yellow
Melting point56°C to 63°C
Appearance (Form)Powder or crystals and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only